4,5,6,7-Tetrahydro-1-benzothiophene-2-sulphonamide
Catalog # APOSOR110986-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:217.308 g/mol
Storage Temperature:Ambient
CAS Number:142294-64-2
Formula:C₈H₁₁NO₂S₂
MDL Number:MFCD25958940