2-(4-Chlorophenoxy)-2-methylpropan-1-ol
Leverancier: Apollo Scientific
Ga naar:
Over dit artikel
- CAS-nummer:14200-96-5
- Formule:C₁₀H₁₃ClO₂
- MDL-nummer:MFCD16304933
- Molecular Weight:200.662 g/mol
- Temperatuur bij opslag:Ambient
2 Opties Hieronder Beschikbaar
Product Details & Documents
- CAS-nummer:14200-96-5
- Formule:C₁₀H₁₃ClO₂
- MDL-nummer:MFCD16304933
- Molecular Weight:200.662 g/mol
- Temperatuur bij opslag:Ambient
Product Family Options
Productinformatie
Aanbevelingen
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



