Order Entry
Netherlands
ContactUsLinkComponent
 
1-(2-Naphthylmethyl)-1H-pyrazol-4-amine
Catalog # APOSOR110929-1G
Supplier:  Apollo Scientific
CAS Number:  
1-(2-Naphthylmethyl)-1H-pyrazol-4-amine
Catalog # APOSOR110929-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :223.273 g/mol
Storage Temperature:Ambient
CAS Number:1240567-95-6
Formula:C₁₄H₁₃N₃
MDL Number:MFCD16810919