Methyl 4-[3-(dimethylamino)prop-2-enoyl]benzoate
Catalog # APOSOR110918-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:233.263 g/mol
Storage Temperature:Ambient
CAS Number:114431-72-0
Formula:C₁₃H₁₅NO₃
MDL Number:MFCD09908108