Order Entry
Netherlands
ContactUsLinkComponent
1-[(4-Methyl-1,3-thiazol-5-yl)sulphonyl]piperidine-4-carboxylic acid
1-[(4-Methyl-1,3-thiazol-5-yl)sulphonyl]piperidine-4-carboxylic acid
Catalog # APOSOR110879-1G
Supplier:  Apollo Scientific
CAS Number:  
1-[(4-Methyl-1,3-thiazol-5-yl)sulphonyl]piperidine-4-carboxylic acid
Catalog # APOSOR110879-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :290.359 g/mol
Storage Temperature:Ambient
CAS Number:1858255-45-4
Formula:C₁₀H₁₄N₂O₄S₂
MDL Number:MFCD26936323