4-[2-{4-(tert-Butyloxycarbonyl)piperazin-1-yl}ethoxy]benzoic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1216502-34-9
:
Formula:C₁₈H₂₆N₂O₅
MDL Number:MFCD11100033
:350.409 g/mol
Storage Temperature:Ambient
Frequently Bought Together