
5-Chlorobenzo[b]thiophene-2-carbohydrazide
Catalog # APOSOR110430-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Verpakking:Bottle
- VPE:1 g
- Seq:3047
Specifications
About this item
:
:226.683 g/mol
:GHS07
Storage Temperature:Kamertemperatuur
CAS Number:87999-20-0
Formula:C₉H₇ClN₂OS
MDL Number:MFCD12032265