
5-Chloro-4-(chloromethyl)-1,2,3-thiadiazole
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:88127-85-9
:
Formula:C₃H₂Cl₂N₂S
MDL Number:MFCD00114816
:169.032 g/mol
Melting Pt:34 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together