
2-(4-Chlorophenyl)piperidine hydrochloride
Catalog # APOSOR1006-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:232.15 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1172776-65-6
Formula:C₁₁H₁₄ClN·ClH
MDL Number:MFCD02178960