Diethoxy-phosphorylethyl-PEG5-ethylphosphonic acid
Catalog # APOSBIPG1380-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:466.398 g/mol
Storage Temperature:Ambient
Formula:C₁₆H₃₆O₁₁P₂
MDL Number:MFCD25424119