
(R)-allylglycinol hydrochloride
Catalog # APOSBICR116-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:137.608 g/mol
:GHS07
Storage Temperature:Freezer
CAS Number:926660-30-2
Formula:C₅H₁₁NO·ClH
MDL Number:MFCD11052925