
6-Deoxy-6-iodo-1,2:3,4-di-O-isopropylidene-α-D-galactopyranose ≥98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:4026-28-2
:
Formula:C₁₂H₁₉IO₅
MDL Number:MFCD00038408
:370.181 g/mol
Melting Pt:56…57 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together