
EMCH.HCl (E-Maleimidocaproic acid hydrazine hydrochloride)
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:175290-73-0
:151038-94-7
Formula:C₁₀H₁₅N₃O₃·ClH
MDL Number:MFCD07363434
:261.705 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together