
LC-SPDP (Succinimidyl 6-[3-(2-pyridylthio)propionamidohexanoate)
Catalog # APOSBICL208-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:425.522 g/mol
Melting Pt:92…94 °C
:GHS07
Storage Temperature:Ambient
CAS Number:158913-22-5
Formula:C₁₈H₂₃N₃O₅S₂
MDL Number:MFCD00083163