
SIAB (N-Succinimidyl (4-iodoacetyl)aminobenzoate)
Catalog # APOSBICL204-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:400.125 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:72252-96-1
Formula:C₁₃H₉IN₂O₅
MDL Number:MFCD00078537