
β-Thionicotinamide adenine dinucleotide, reduced form disodium salt 92+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1921-48-8
:
Formula:C₂₁H₂₇N₇O₁₃P₂S·2Na
MDL Number:MFCD00133854
:725.47 g/mol
:GHS07
Storage Temperature:Freezer
Frequently Bought Together