Order Entry
Netherlands
ContactUsLinkComponent
1,4-Dimethylpyridinium p-toluenesulphonate ≥98%, Sigma-Aldrich®
Irritant
1,4-Dimethylpyridinium p-toluenesulphonate ≥98%, Sigma-Aldrich®
Catalog # 514888-10G
CAS Number:  
undefined
Irritant
1,4-Dimethylpyridinium p-toluenesulphonate ≥98%, Sigma-Aldrich®
Catalog # 514888-10G
CAS Number:  

Specifications

  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

 :
 :279.35 g/mol
Melting Pt:148…151 °C (lit.)
 :GHS07
Storage Temperature:Ambient
CAS Number:78105-28-9
Formula:C₁₄H₁₇NO₃S
MDL Number:MFCD00137110