
Glutaconaldehydedianil hydrochloride, Sigma-Aldrich®
Catalog # 247405-5G
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:284.79 g/mol
Melting Pt:173…175 °C (lit.)
:GHS07
Storage Temperature:Ambient
CAS Number:1497-49-0
Formula:C₁₇H₁₆N₂
MDL Number:MFCD00012630