Order Entry
Netherlands
ContactUsLinkComponent
Tetrabromophenol Blue sodium salt 85% (dye content) ≥84.5% (by HPLC), Sigma-Aldrich®
Irritant
Tetrabromophenol Blue sodium salt 85% (dye content) ≥84.5% (by HPLC), Sigma-Aldrich®
Certificates
undefined
Irritant
Tetrabromophenol Blue sodium salt 85% (dye content) ≥84.5% (by HPLC), Sigma-Aldrich®

Specification Test Results

  • Appearance (Form):
    Powder or Crystals
  • Infrared spectrum:
    Conforms to Structure
  • Purity (HPLC):
    ≥84.5%
  • Wavelength:
    607 - 613 nm
  • Extinction Coefficient:
    ≥55000
  • Extinction Coefficient, C = 0.005 g/L in H2O:
    ≥59000

Specification Test Results

Frequently Bought Together