Biocide PT2, UMONIUM38® MASTER
HUCKPF10110
: HUCKERTS
- Capacity:1 L
- Seq:01
- Packed:Dosing bottles
- Pk:12
UMONIUM38® MASTER combines the multiple functions of a cleaner-degreaser, disinfectant and deodorant for sanitary equipment, furniture, high and low surfaces, clean rooms, and so on.
It is also used to disinfect textiles and PPE (Personal Protective Equipment) either by soaking the fabric in a diluted solution, or directly in a washing machine, instead of softener.
: European standards: EN 1040:2006, EN 1275:2005, EN 1276:2010, EN 1650:2008, EN 14348:2005, EN 14476:2013, EN 13697:2001, EN 14563:2009. Complies with biocide regulation 528/2012. Quality and traceability guaranteed by ISO 9001:2015. AMS 1453 validation.
: Use biocides safely. Always read the label and product information before use.