Doripenem hydrate
Supplier: Thermo Fisher Scientific
About this item
- Bioactive
- CAS Number:1820954-21-9
- Formula:C₁₅H₂₄N₄O₆S₂·xH₂O
- MDL Number:MFCD09749887
- Molecular Weight:420.5 g/mol
- Storage Temperature:Refrigerator
2 Options Available Below
Product Details & Documents
- Bioactive
- CAS Number:1820954-21-9
- Formula:C₁₅H₂₄N₄O₆S₂·xH₂O
- MDL Number:MFCD09749887
- Molecular Weight:420.5 g/mol
- Storage Temperature:Refrigerator
Specifications for Product Family
Specification Test Results
- Appearance:White to beige Powder
- Infrared spectrum:Conforms
- Water:≤6 %
- HPLC:≥97.5 % (on dry substance)
- Residue after ignition:≤0.1 %
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



