Hard hat integrated safety glasses
Catalog # MMMAV9C
Supplier: 3M
Specifications
- Description:Hard hat integrated safety glasses V9C
- Environmentally Preferable:
- Pk:1
Specifications
About this item
Hard hat integrated safety glasses, V9C, are clear lens safety glasses designed to attach to 3M hard hats for protection from particles and UV.
- Provides eye protection from particles and UV
- Easily attaches to 3M hard hats
- Durable anti-scratch and anti-fog lens coating helps minimise physical damage and fogging
- Easy to adjust for a good fit
- Faceshield marking 2C-1,2 3M 1 F
Hard hat integrated safety glasses, V9C, feature a unique design that allows them to slide easily between the hard hat shell and headband, fitting closely to the wearer’s cheeks to provide protection.
Certifications: EN 166:2001