Analytical standard, Cyprodinil, reference material, Fluka™
Catalog # HONCR1720-100MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mg
- Standards:Reference material
- CAS No.:121552-61-2
- Description:Cyprodinil
- Description:Cyprodinil
- Form:Neat