Analytical standard, Prochloraz, reference material, Fluka™
Catalog # HONCR1788-100MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mg
- Standards:Reference material
- CAS No.:67747-09-5
- Description:Prochloraz
- Description:Prochloraz
- Form:Neat