Analytical standard, Cymoxanil, reference material, Fluka™
Catalog # HONCR1785-100MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mg
- Standards:Reference material
- CAS No.:57966-95-7
- Description:Cymoxanil
- Formula:C₇H₁₀N₄O₃
- Storage temperature:Ambient
- Description:Cymoxanil
- Form:Neat