Order Entry
Netherlands
ContactUsLinkComponent
Kerosene, Reagent Grade low odor, Riedel-de Haën™
Kerosene, Reagent Grade low odor, Riedel-de Haën™
Catalog # 329460-18.9L
CAS Number:  
Kerosene, Reagent Grade low odor, Riedel-de Haën™
Catalog # 329460-18.9L
CAS Number:  

Specifications

  • Pack type:
    Stainless steel can
  • Pk:
    18,9 L
  • Seq:
    5277
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Identity (IR):
    complying
  • Autoignition Temperature:
    442 °F
  • Explosion Limit:
    5 %
  • Vapor Pressure:
    0.23 mmHg ( 20 °C)

Specification Test Results

About this item

 :
Boiling Pt:175…325 °C
Density:0.8 g/cm3 (20 °C)
Flash Pt:38 °C
Storage Temperature:Ambient
EINECS:232-366-4
UN:1223
ADR:3,III
 :GHS02
 :GHS08
 :GHS07
 :GHS09
CAS Number:8008-20-6
MDL Number:MFCD00135561