Order Entry
Netherlands
ContactUsLinkComponent
Cable ties, stainless steel
Cable ties, stainless steel
Catalog # BRDY134057
Supplier:  Brady
undefined
Cable ties, stainless steel
Catalog # BRDY134057
Supplier:  Brady
Specifications
  • Colour:
    Silver
  • Description:
    Cable ties
  • Dimensions:
    4,5×127 mm
  • Pk:
    100
Specifications
About this item
Brady B-7316 stainless steel cable ties are made from 316 grade stainless steel and are designed to withstand harsh environments, and offer excellent outdoor durability.
  • Thickness of 0,5 mm