Order Entry
Netherlands
ContactUsLinkComponent
N-Ethyldiisopropylamine ≥98%, GPR RECTAPUR®
Flammable
Toxic
Corrosive
N-Ethyldiisopropylamine ≥98%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
Flammable
Toxic
Corrosive
N-Ethyldiisopropylamine ≥98%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 98 %
  • Density (20/4):
    0.755 --> 0.758
  • Water:
    Max. 0.2 %

Specification Test Results

Frequently Bought Together