
Platinum(II) acetylacetonate for synthesis, Sigma-Aldrich®
8.14689.0001
: Merck
:
- Verpakking:Glass bottle
- VPE:1 g
- Seq:3047
- Solubility:(20 °C) insoluble
- Melting Point:250 - 252 °C
- Molar Mass:393.3 g/mol
:
:393.3 g/mol
:250…252 °C
:∼1 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
:239-223-5
:GHS07
:15170-57-7
:C₁₀H₁₄O₄Pt
:MFCD00000028