Order Entry
Netherlands
ContactUsLinkComponent
Platinum(II) acetylacetonate 98%
Irritant
Health
Platinum(II) acetylacetonate 98%
Certificates
undefined
Irritant
Health
Platinum(II) acetylacetonate 98%

Specification Test Results

  • Appearance:
    Yellow to yellow-green Powder
  • Infrared spectrum:
    Conforms
  • Assay:
    ≥97.5 % (ex Pt)

Specification Test Results

Related Information

Frequently Bought Together