Manganese(II) sulphate
Catalog # HACH107166
Supplier: Hach
CAS Number:
Specifications
- Pk:50
- Seq:0050
- Environmentally Preferable:
Specifications
About this item
Synonyms: Manganese sulphate, Manganous sulphate
:
:151 g/mol
EINECS:232-089-9
UN:3077
:GHS08
:GHS09
CAS Number:7785-87-7
Formula:MnO₄S
MDL Number:MFCD00003464
Melting Pt:700…720 °C
Density:3.25 g/cm³ (20 °C)
ADR:9 III
Merck Index:12,05782