


Molybdenum(VI) oxide ≥99.998% (metals basis excluding W), Puratronic® (max. 300 ppm W)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1313-27-5
:
Formula:MoO₃
MDL Number:MFCD00003469
:143.94 g/mol
Merck Index:14,6239
Boiling Pt:1155 °C (subl.)
Melting Pt:795 °C
Density:4.692
Storage Temperature:Ambient
EINECS:215-204-7
UN:3288
ADR:6.1,III
:GHS06
:GHS08
:GHS07
Frequently Bought Together