
Tetrapentylammonium iodide ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2498-20-6
:
Formula:C₂₀H₄₄IN
MDL Number:MFCD00041980
:425.48 g/mol
Melting Pt:135…139 °C
Density:1.18 g/cm³ (25 °C)
EINECS:219-688-0
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together