Cetrimonium chloride
Catalog # MOLE53473813-100G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:320 g/mol
Melting Pt:232…233 °C
Density:0,9 g/cm³ (20 °C)
EINECS:203-928-6
CAS Number:112-02-7
Formula:C₁₉H₄₂ClN
MDL Number:MFCD00011773