1-Iodo-3,5-dinitrobenzene 97%, Sigma-Aldrich®
452769-1G
:
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
- Environmentally Preferable:
- Appearance (Colour):Conforms to Requirements
Faint to Dark Yellow - Appearance (Form):Conforms to Requirements
Powder or Crystals - Infrared spectrum:Conforms to Structure
- Carbon:48.2 - 51.8%
- Nitrogen:14.0 - 15.1%
- Purity (HPLC):≥96.5%
:
:294.01 g/mol
:GHS07
:Room Temperature
:6276-04-6
:C₆H₃IN₂O₄
:MFCD00024330
:213...215 °C (lit.)
:M-Clarity™ quality level = MQ200