Zofenopril Calcium ≥97.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:81938-43-4
:MFCD11858294
Formula:C₄₄H₄₄CaN₂O₈S₄
MDL Number:MFCD08704648
:897.17 g/mol
Merck Index:14,10186
Melting Pt:246°C
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 97.0 area%
Specification Test Results
Frequently Bought Together