
Xylemin dihydrochloride ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:89690-11-9
:
Formula:C₇H₁₈N₂·₂HCl
:203.15 g/mol
Melting Pt:272°C
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Argentmetric Titration):min. 98.0 %
- Purity (with Total Nitrogen):min. 96.0 %
Specification Test Results
Frequently Bought Together