(R)-(+)-XylBINAP ≥97.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:137219-86-4
:
Formula:C₅₂H₄₈P₂
MDL Number:MFCD01630821
:734.90 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 97.0 area%
- Optical purity (LC):min. 99.0 ee%
Specification Test Results
Frequently Bought Together