Xanthohumol ≥97.0% (by HPLC)
Catalog # TCIAX0068-25MG
Supplier: TCI
CAS Number:
Specifications
- Pk:25 mg
- Seq:2315
Specifications
Specification Test Results
- Appearance:Light yellow to Brown powder to crystal
- Purity (HPLC):min. 97.0 area%
- Melting point:158.0 to 163.0 deg-C
Specification Test Results
About this item
:
:354.40 g/mol
Storage Temperature:Refrigerator
CAS Number:6754-58-1
Formula:C₂₁H₂₂O₅
MDL Number:MFCD00210576