
1-Vinylimidazole bis(trifluoromethanesulphonyl)imide ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAV0145-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White or Colorless to Yellow to Orange powder to lump to clear liquid
- Purity (HPLC):min. 98.0 area%
- Purity (Ion exchange titration):min. 98.0 %
- Melting point:35.0 to 39.0 deg-C
Specification Test Results
About this item
:
:375.26 g/mol
:GHS07
CAS Number:1013027-27-4
Formula:C₇H₇F₆N₃O₄S₂