
3-Vinylphenylboronic acid (contains varying amounts of Anhydride)
: TCI
:15016-43-0
:
:C₈H₉BO₂
:MFCD01074679
:147.97 g/mol
:145°C
:GHS07
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):96.0 to 114.0 %
Frequently Bought Together