Order Entry
Netherlands
ContactUsLinkComponent
Violanthrone-79 ≥95.0% (by HPLC)
Violanthrone-79 ≥95.0% (by HPLC)
Supplier:  TCI
Certificates
undefined
Violanthrone-79 ≥95.0% (by HPLC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Orange to Brown to Dark purple powder to crystal
  • Purity (HPLC):
    min. 95.0 area%

Specification Test Results

Frequently Bought Together