Order Entry
Netherlands
ContactUsLinkComponent
 
DL-Valine methyl ester hydrochloride ≥98.0% (by titrimetric analysis)
 :  TCI
 
undefined
DL-Valine methyl ester hydrochloride ≥98.0% (by titrimetric analysis)
 :  TCI

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 95.0 area%
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Melting point:
    103.0 to 107.0 deg-C

 

Frequently Bought Together