
DL-Valinol ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:16369-05-4
:
Formula:C₅H₁₃NO
MDL Number:MFCD00004730
:103.17 g/mol
Boiling Pt:87°C
Flash Pt:90°C
EINECS:240-425-0
:GHS07
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together