L-Valine-tert-butyl ester hydrochloride ≥98.0% (by total nitrogen and titration analysis)
TCIAV0097-5G
: TCI
:
- VPE:5 g
- Seq:3173
- Appearance:White to Almost white powder to crystal
- Purity (with Total Nitrogen):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Specific rotation [a]20/D:+4.0 to +6.0 deg (C=2、H2O)
:
:209.7 g/mol
:13518-40-6
:C₉H₁₉NO₂·HCl
:MFCD00034863