N-Succinimidyl [(tert-Butoxycarbonyl)aminooxy]acetate ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI
Certificates
About this item
CAS Number:80366-85-4
:
Formula:C₁₁H₁₆N₂O₇
MDL Number:MFCD18382162
:288.26 g/mol
Storage Temperature:Freezer
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:112.0 to 116.0 deg-C
Specification Test Results
Frequently Bought Together