Order Entry
Netherlands
ContactUsLinkComponent
 
Undecyl ether ≥97.0% (by GC)
  TCIAU0051-25G
 :  TCI
 :  
Undecyl ether ≥97.0% (by GC)
  TCIAU0051-25G
 :  TCI
 :  

 

  • Pk:
    25 g
  • Seq:
    3308

 

 

  • Appearance:
    White or Colorless to Light yellow powder to lump to clear liquid
  • Purity (GC):
    min. 97.0 %

 

 

 :
 :326.61 g/mol
 :256-117-4
 :43146-97-0
 :C₂₂H₄₆O
 :MFCD00059282