N,N,N',N'-Tetrakis[4-(dibutylamino)phenyl]-1,4-phenylenediamine ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:4182-80-3
:
Formula:C₆₂H₉₂N₆
MDL Number:MFCD09832942
:921.46 g/mol
EINECS:224-057-8
Specification Test Results
- Appearance:Light yellow to Amber to Dark green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 97.0 %
- Melting point:103.0 to 107.0 deg-C
Specification Test Results
Frequently Bought Together