
2,3,5-Trichlorophenylboronic acid ≥98.0% (by HPLC, titration analysis) (contains varying amounts of Anhydride)
Supplier: TCI
Certificates
About this item
CAS Number:212779-19-6
:
Formula:C₆H₄BCl₃O₂
MDL Number:MFCD01630856
:225.26 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):98.0 to 108.0 %
Specification Test Results
Frequently Bought Together