
3-(Trimethoxysilyl)propyl 2-Bromo-2-methylpropanoate ≥96.0% (by GC)
Catalog # TCIAT3845-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 96.0 %
Specification Test Results
About this item
:
:329.26 g/mol
Boiling Pt:115°C
:GHS07
CAS Number:314021-97-1
Formula:C₁₀H₂₁BrO₅Si
MDL Number:MFCD09953896